ADVH0430AC94 AChemBlock

4'-formylbiphenyl-4-carboxylic acid

  • MDL number MFCD03424613
  • Cas Number : 70916-98-2
  • Molecular Weight : 226.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-formylbiphenyl-4-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30785
Availability details for
Quantity
  • Availability
  • Add to Cart