ADVH0430ACD5 AChemBlock

3-(2-fluorophenyl)-3-oxopropanenitrile

  • MDL number MFCD02260800
  • Cas Number : 31915-26-1
  • Molecular Weight : 163.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-2-fluorophenyl-3-oxopropanenitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30850
Availability details for
Quantity
  • Availability
  • Add to Cart