ADVH0430ACE0 AChemBlock

methyl 1-methyl-1H-imidazole-4-carboxylate

  • MDL number MFCD02179561
  • Cas Number : 17289-19-9
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-methyl-1h-imidazole-4-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30861
Availability details for
Quantity
  • Availability
  • Add to Cart