ADVH0430ACE2 AChemBlock

4-(4-fluorophenyl)butanoic acid

  • MDL number MFCD03788503
  • Cas Number : 589-06-0
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-4-fluorophenyl-butanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30863
Availability details for
Quantity
  • Availability
  • Add to Cart