ADVH0430ACE9 AChemBlock

tert-Butyl 3-hydroxypropylmethylcarbamate

  • MDL number MFCD09751790
  • Cas Number : 98642-44-5
  • Molecular Weight : 189.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-hydroxypropylmethylcarbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30870
Availability details for
Quantity
  • Availability
  • Add to Cart