ADVH0430ACEF AChemBlock

2-amino-4-chlorothiazole-5-carbaldehyde

  • MDL number MFCD00204993
  • Cas Number : 76874-79-8
  • Molecular Weight : 162.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-amino-4-chlorothiazole-5-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30876
Availability details for
Quantity
  • Availability
  • Add to Cart