ADVH0430AD08 AChemBlock

Methyl 3,5-difluoropyridine-2-carboxylate

  • MDL number MFCD11616886
  • Cas Number : 955885-64-0
  • Molecular Weight : 173.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-5-difluoropyridine-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30901
Availability details for
Quantity
  • Availability
  • Add to Cart