ADVH0430ADAC AChemBlock

(4-methoxypyridin-2-yl)methanamine dihydrochloride

  • MDL number MFCD20327973
  • Cas Number : 1344046-06-5
  • Molecular Weight : 211.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-methoxypyridin-2-yl-methanamine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31065
Availability details for
Quantity
  • Availability
  • Add to Cart