ADVH0430AE06 AChemBlock

quinoline-4-carbonitrile

  • MDL number MFCD00972111
  • Cas Number : 2973-27-5
  • Molecular Weight : 154.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/quinoline-4-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31155
Availability details for
Quantity
  • Availability
  • Add to Cart