ADVH0430AEAD AChemBlock

5-bromo-7-methoxy-1H-indazole

  • MDL number MFCD11878570
  • Cas Number : 1374652-62-6
  • Molecular Weight : 227.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-7-methoxy-1h-indazole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31322
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart