ADVH0430AFA5 AChemBlock

hex-5-yn-1-amine hydrochloride

  • MDL number MFCD18839147
  • Cas Number : 102169-54-0
  • Molecular Weight : 133.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/hex-5-yn-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31570
Availability details for
Quantity
  • Availability
  • Add to Cart