ADVH0430B0E0 AChemBlock

1-(1-methyl-1H-pyrazol-4-yl)ethanone

  • MDL number MFCD00159640
  • Cas Number : 37687-18-6
  • Molecular Weight : 124.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-1-methyl-1h-pyrazol-4-yl-ethanone.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31882
Availability details for
Quantity
  • Availability
  • Add to Cart