ADVH0430B213 AChemBlock

3-(trifluoromethyl)pyridine-2-sulfonyl chloride

  • MDL number MFCD16093662
  • Cas Number : 104040-75-7
  • Molecular Weight : 245.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-trifluoromethyl-pyridine-2-sulfonyl-chloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32189
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart