ADVH0430B3C2 AChemBlock

5-(Tetrahydro-2H-pyran-4-yl)-1H-pyrazol-3-amine

  • MDL number MFCD20502055
  • Cas Number : 1000896-69-4
  • Molecular Weight : 167.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-tetrahydro-2h-pyran-4-yl-1h-pyrazol-3-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32623
Availability details for
Quantity
  • Availability
  • Add to Cart