ADVH0430B428 AChemBlock

Pyrrolidine-1-sulfonamide

  • MDL number MFCD11171672
  • Cas Number : 4108-88-7
  • Molecular Weight : 150.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/pyrrolidine-1-sulfonamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O32725
Availability details for
Quantity
  • Availability
  • Add to Cart