ADVH0430B7FA AChemBlock

Fluonid

  • MDL number N/A
  • Cas Number : 790-69-2
  • Molecular Weight : 315.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://www.achemblock.com/fluonid.html

Safety Information

msds https://www.achemblock.com/AC_SDS.php?Catalog=O33703
Availability details for
Quantity
  • Availability
  • Add to Cart