ADVH0430B9D9 AChemBlock

3-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)Benzenamine

  • MDL number MFCD22494808
  • Cas Number : 1350377-66-0
  • Molecular Weight : 249.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-methoxy-4-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-benzenamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O34165
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart