ADVH0430BB92 AChemBlock

(1-Cyclopropylethyl)amine hydrochloride

  • MDL number MFCD06685893
  • Cas Number : 42390-64-7
  • Molecular Weight : 121.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-cyclopropylethyl-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-640-HCl
Availability details for
Quantity
  • Availability
  • Add to Cart