ADVH14DBAF46 AChemBlock

6-Aza-spiro[3.4]octane

  • MDL number MFCD09759054
  • Cas Number : 765-64-0
  • Molecular Weight : 111.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form White solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-aza-spiro-3-4-octane-oxalate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6679
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart