ADVH14DBAF4A AChemBlock

1-(tert-butyl) 2-methyl (2S,4S)-4-methoxypyrrolidine-1,2-dicarboxylate

  • MDL number MFCD16621201
  • Cas Number : 215918-38-0
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-2-pyrrolidinedicarboxylic-acid-4-methoxy-1-1-1-dimethylethyl-2-methyl-ester-2s-4s.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14100
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart