ADVH14DBAF5D AChemBlock

3-Methyl-D-phenylalanine

  • MDL number MFCD01860871
  • Cas Number : 114926-39-5
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-methyl-d-phenylalanine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42532
Availability details for
Quantity
  • Availability
  • Add to Cart