ADVH14DBAF7B AChemBlock

Fmoc-2-Methyl-D-phenylalanine

  • MDL number MFCD00671404
  • Cas Number : 352351-63-4
  • Molecular Weight : 401.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/fmoc-2-methyl-d-phenylalanine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38612
Availability details for
Quantity
  • Availability
  • Add to Cart