ADVH14DBAF83 AChemBlock

Fmoc-3,4-Dichloro-L-phenylalanine

  • MDL number MFCD00273473
  • Cas Number : 177966-59-5
  • Molecular Weight : 456.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/fmoc-3-4-dichloro-l-phenylalanine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40453
Availability details for
Quantity
  • Availability
  • Add to Cart