ADVH14DBAF8D AChemBlock

4-Cyano-DL-phenylalanine

  • MDL number MFCD01862353
  • Cas Number : 22888-47-7
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-cyano-dl-phenylalanine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39528
Availability details for
Quantity
  • Availability
  • Add to Cart