ADVH14DBAFB7 AChemBlock

3-Amino-3-(2-naphthyl)propionic acid

  • MDL number MFCD01871322
  • Cas Number : 129042-57-5
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-amino-3-2-naphthyl-propionic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41805
Availability details for
Quantity
  • Availability
  • Add to Cart