ADVH14DBAFC0 AChemBlock

Boc-3-Amino-3-(4-bromophenyl)propionic acid

  • MDL number MFCD02093998
  • Cas Number : 282524-86-1
  • Molecular Weight : 344.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/boc-3-amino-3-4-bromophenyl-propionic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39075
Availability details for
Quantity
  • Availability
  • Add to Cart