ADVH14DBAFC1 AChemBlock

Boc-3-Amino-3-(2-methoxyphenyl)propionic acid

  • MDL number MFCD02090692
  • Cas Number : 284493-52-3
  • Molecular Weight : 295.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/boc-3-amino-3-2-methoxyphenyl-propionic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39067
Availability details for
Quantity
  • Availability
  • Add to Cart