ADVH14DBB0BA AChemBlock

(R)-2-Amino-2-(3-trifluoromethylphenyl)ethanol

  • MDL number MFCD09253707
  • Cas Number : 1035490-73-3
  • Molecular Weight : 205.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-2-amino-2-3-trifluoromethylphenyl-ethanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42914
Availability details for
Quantity
  • Availability
  • Add to Cart