ADVH14DBB0E0 AChemBlock

(R)-N-Cbz-2-amino-1-phenylethylamine

  • MDL number MFCD09260583
  • Cas Number : 130406-35-8
  • Molecular Weight : 270.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-n-cbz-2-amino-1-phenylethylamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41757
Availability details for
Quantity
  • Availability
  • Add to Cart