ADVH14DBB12D AChemBlock

(S)-b-Amino-4-methoxybenzenepropanol

  • MDL number MFCD17214628
  • Cas Number : 20989-19-9
  • Molecular Weight : 181.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-b-amino-4-methoxybenzenepropanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39832
Availability details for
Quantity
  • Availability
  • Add to Cart