ADVH14DBB162 AChemBlock

(S)-1-Phenyl-3-(pyrrolidin-1-yl)propan-2-amine dihydrochloride

  • MDL number MFCD31560215
  • Cas Number : 200267-75-0
  • Molecular Weight : 277.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-1-phenyl-3-pyrrolidin-1-yl-propan-2-amine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40015
Availability details for
Quantity
  • Availability
  • Add to Cart