ADVH14DBB16F AChemBlock

S-N1,N1-Dimethyl-3-phenylpropane-1,2-diamine dihydrochloride

  • MDL number MFCD21607233
  • Cas Number : 29802-26-4
  • Molecular Weight : 251.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-n1-n1-dimethyl-3-phenylpropane-1-2-diamine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38986
Availability details for
Quantity
  • Availability
  • Add to Cart