ADVH14DBB179 AChemBlock

N1,N1,3-Trimethyl-1,2-butanediamine dihydrochloride

  • MDL number MFCD29046379
  • Cas Number : 773045-76-4
  • Molecular Weight : 203.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n1-n1-3-trimethyl-1-2-butanediamine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36129
Availability details for
Quantity
  • Availability
  • Add to Cart