ADVH14DBB1B7 AChemBlock

1-(3-Piperidinylcarbonyl)-pyrrolidine hydrochloride

  • MDL number MFCD11520873
  • Cas Number : 35090-94-9
  • Molecular Weight : 218.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-piperidinylcarbonyl-pyrrolidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38629
Availability details for
Quantity
  • Availability
  • Add to Cart