ADVH14DBB1E1 AChemBlock

4-Cyano-4-(3-methylphenyl)cyclohexanone

  • MDL number MFCD11849771
  • Cas Number : 73312-15-9
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-cyano-4-3-methylphenyl-cyclohexanone.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36377
Availability details for
Quantity
  • Availability
  • Add to Cart