ADVH14DBB1E9 AChemBlock

(3,4-Dichlorophenyl)-4-piperidinyl-methanone hydrochloride

  • MDL number MFCD08544118
  • Cas Number : 269731-80-8
  • Molecular Weight : 294.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-4-dichlorophenyl-4-piperidinyl-methanone-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39176
Availability details for
Quantity
  • Availability
  • Add to Cart