ADVH14DBB216 AChemBlock

N-(3-Fluorophenylmethyl)isopropylamine

  • MDL number MFCD08060644
  • Cas Number : 90389-87-0
  • Molecular Weight : 167.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-3-fluorophenylmethyl-isopropylamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35255
Availability details for
Quantity
  • Availability
  • Add to Cart