ADVH14DBB223 AChemBlock

N-(4-Methylphenylmethyl)tert-butylamine

  • MDL number MFCD01534063
  • Cas Number : 55980-45-5
  • Molecular Weight : 177.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-4-methylphenylmethyl-tert-butylamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37416
Availability details for
Quantity
  • Availability
  • Add to Cart