ADVH14DBB233 AChemBlock

2-Amino-4'-hydroxy-acetophenone hydrochloride

  • MDL number MFCD08436951
  • Cas Number : 19745-72-3
  • Molecular Weight : 187.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-amino-4-hydroxy-acetophenone-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40081
Availability details for
Quantity
  • Availability
  • Add to Cart