ADVH14DBB265 AChemBlock

1-(3-Bromophenyl)-cyclobutanemethanamine

  • MDL number MFCD11642693
  • Cas Number : 915690-61-8
  • Molecular Weight : 240.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-bromophenyl-cyclobutanemethanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35134
Availability details for
Quantity
  • Availability
  • Add to Cart