ADVH14DBB27E AChemBlock

a-Cyclopropyl-4-fluoro-benzylamine

  • MDL number MFCD05191953
  • Cas Number : 705-14-6
  • Molecular Weight : 165.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/a-cyclopropyl-4-fluoro-benzylamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36527
Availability details for
Quantity
  • Availability
  • Add to Cart