ADVH14DBB2C2 AChemBlock

4'-(Aminomethyl)-biphenyl-4-ol hydrochloride

  • MDL number MFCD09054666
  • Cas Number : 60277-40-9
  • Molecular Weight : 235.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-aminomethyl-biphenyl-4-ol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37156
Availability details for
Quantity
  • Availability
  • Add to Cart