ADVH14DBB2D5 AChemBlock

3-Nitro-phenethylamine hydrochloride

  • MDL number MFCD08059237
  • Cas Number : 19008-62-9
  • Molecular Weight : 202.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-nitro-phenethylamine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40197
Availability details for
Quantity
  • Availability
  • Add to Cart