ADVH14DBB2D6 AChemBlock

2-Nitro-phenethylamine hydrochloride

  • MDL number MFCD07371521
  • Cas Number : 861337-74-8
  • Molecular Weight : 202.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-nitro-phenethylamine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35716
Availability details for
Quantity
  • Availability
  • Add to Cart