ADVH14DBB2E0 AChemBlock

1-[3-(Trifluoromethyl)phenyl]-cyclopropanamine

  • MDL number MFCD07374480
  • Cas Number : 503417-34-3
  • Molecular Weight : 201.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-trifluoromethyl-phenyl-cyclopropanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37812
Availability details for
Quantity
  • Availability
  • Add to Cart