ADVH14DBB2E2 AChemBlock

1-(3-Methylphenyl)-cyclopropanamine

  • MDL number MFCD07374481
  • Cas Number : 503417-30-9
  • Molecular Weight : 147.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-methylphenyl-cyclopropanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37815
Availability details for
Quantity
  • Availability
  • Add to Cart