ADVH14DBB2E7 AChemBlock

1-(4-Bromophenyl)-cyclopropanamine

  • MDL number MFCD07374473
  • Cas Number : 345965-54-0
  • Molecular Weight : 212.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-4-bromophenyl-cyclopropanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38660
Availability details for
Quantity
  • Availability
  • Add to Cart