ADVH14DBB2F4 AChemBlock

2-Amino-N-methyl-acetamide hydrochloride

  • MDL number MFCD00153440
  • Cas Number : 49755-94-4
  • Molecular Weight : 124.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-amino-n-methyl-acetamide-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37877
Availability details for
Quantity
  • Availability
  • Add to Cart