ADVH14DBB307 AChemBlock

2-Chloro-N-methyl-benzeneethanamine

  • MDL number MFCD11212391
  • Cas Number : 52516-17-3
  • Molecular Weight : 169.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-n-methyl-benzeneethanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37635
Availability details for
Quantity
  • Availability
  • Add to Cart