ADVH14DBB30F AChemBlock

1-(3-Chlorophenyl)ethylamine

  • MDL number MFCD05215231
  • Cas Number : 24358-43-8
  • Molecular Weight : 155.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-chlorophenyl-ethylamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39424
Availability details for
Quantity
  • Availability
  • Add to Cart